Kalcium standardlösning, 1000 mg/L Ca in 5% salpetersyra (av CaCO3) Specpure® standard för AAS

Leverantör: Thermo Fisher Scientific

88057.AE 88057.AP
88057.AEEA 970 SEK
88057.AE 88057.AP
Kalcium standardlösning, 1000 mg/L Ca in 5% salpetersyra (av CaCO3) Specpure® standard för AAS
Standarder Cationic Standards Calcium Standards

Molecular Formula: CaCO₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085314


Certifikat: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR