Silvernitrat 0.025 mol/l (0.025 N)

Leverantör: VWR Chemicals
Fara

Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt

88017.320EA 1260 SEK
88017.320
Silvernitrat 0.025 mol/l (0.025 N)
Silvernitrat
Formel: AgNO₃
MW: 169.87 g/mol
Kokpunkt: 444
Smältpunkt: 210
Storage Temperature: Omgivande
MDL Number: MFCD00003414
CAS nummer: 7761-88-8
EINECS: 231-853-9
UN: 1993
ADR: 3 II
Merck Index: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR