Polyethylene glycol Standard ReadyCal Set Mp 102-40′000, for GPC
Leverantör: Merck
87976-1SET-F
87976-1SET-FEA 6410 SEK
87976-1SET-F
Polyethylene glycol Standard ReadyCal Set Mp 102-40′000, for GPC
Standarder Standarder för kromatografi
Set contains 12 different polyethyleneglycol standards with Mp ~106, ~330, ~420, ~600, ~1500, ~2010, ~3120, ~6240, ~8600, ~12000, ~23000, ~40000; always 4 standards are lyophilised in a 1,5 ml vial (green, red or white); for 1 complete calibration use one of each of the green, red and white vials; 1 set contains 30 vials for 10 complete calibrations
Certifikat: Exact values can be taken from the accompanying certificate of analysis.
Observera: Storage temperature: 2 to 8 °C. Limited shelf life, expiry date on the label.