Potassium heptaiodobismuthate 8% aqueous L(+)-tartaric acid solution with potassium iodide Reag. Ph. Eur. 1070602

Leverantör: VWR Chemicals

87914.180EA 2425 SEK
87914.180
Potassium heptaiodobismuthate 8% aqueous L(+)-tartaric acid solution with potassium iodide Reag. Ph. Eur. 1070602
Potassium heptaiodobismuthate

Beställningsinformation: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formel: BiI₇K₄
MW: 1253.71 g/mol
CAS nummer: 41944-01-8
EINECS: 255-595-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR