Saltsyra 0,25% methanol Reag. Ph. Eur. 1053203

Leverantör: VWR Chemicals
Fara

Synonyms: Hydrochloric acid solution, Muriatic acid

87868.180EA 504 SEK
87868.180
Saltsyra 0,25% methanol Reag. Ph. Eur. 1053203
Saltsyra

Beställningsinformation: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formel: HCl
MW: 36.46 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00011324
CAS nummer: 7647-01-0
EINECS: 231-595-7
UN: 1993
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR