Järn(III)klorid 105 g/l aqueous solution Reag. Ph. Eur. 1037801

Leverantör: VWR Chemicals
Fara

Synonyms: Iron trichloride, Ferric chloride

87833.290EA 930 SEK
87833.290
Järn(III)klorid 105 g/l aqueous solution Reag. Ph. Eur. 1037801
Järn(III)klorid

Beställningsinformation: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formel: FeCl₃
MW: 162.21 g/mol
Kokpunkt: 315
Smältpunkt: 260
MDL Number: MFCD00011005
CAS nummer: 7705-08-0
EINECS: 231-729-4
UN: 2582
ADR: 8 III
Merck Index: 13,04048

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR