Vanadinpentoxid solution_SC sulphuric acid Reag. Ph. Eur. 1034001

Leverantör: VWR Chemicals

Synonyms: Vanadium pentoxide, di-Vanadium pentaoxide

87828.180EA 1395 SEK
87828.180
Vanadinpentoxid solution_SC sulphuric acid Reag. Ph. Eur. 1034001
Vanadinpentoxid

Beställningsinformation: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formel: O₅V₂
MW: 181.88 g/mol
Kokpunkt: 1750
Smältpunkt: 690
MDL Number: MFCD00011457
CAS nummer: 1314-62-1
Merck Index: 13,09987

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR