Metanol-D4 Isotope_(99.8% D) NMR

Leverantör: VWR Chemicals
Fara

Synonyms: Methanol-[²H₄], Hydroxy methane-D4, Methanol-d4, Methyl-D3 alcohol-D, Carbinol-D4, Wood alcohol-D4, Methyl alcohol-D4, Tetradeuteromethanol

87157.0010EA 1450 SEK
87157.0010 87157.0100 87157.0025 87157.0006 87157.0011
Metanol-D4 Isotope_(99.8% D) NMR
Metanol-D4
Formel: D₃COD
MW: 36.01 g/mol
Kokpunkt: 65.3 °C (1013 hPa)
Smältpunkt: –99 °C
Densitet: 0.888 g/cm³ (20 °C)
Flampunkt: 11 °C
Storage Temperature: Omgivande
MDL Number: MFCD00044637
CAS nummer: 811-98-3
EINECS: 212-378-6
UN: 1230
ADR: 3 (6.1) II

Order Now

Specifikation testresultat

Assay (on anhydrous substance) Min. 99.9 %
Isotopic enrichment (FT NMR 400 MHz)(D) Min. 99.80 %
Water (HDO+D2O) Max. 0.03 %

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR