1,2-Diklorbenzen ≥99%, AnalaR NORMAPUR®

Leverantör: VWR Chemicals
Varning

Synonyms: o-Diklorbenzen

87064.290EA 1125 SEK
87064.290 87064.320
1,2-Diklorbenzen ≥99%, AnalaR NORMAPUR®
1,2-Diklorbenzen
Formel: C₆H₄Cl₂
MW: 147 g/mol
Kokpunkt: 180,5
Smältpunkt: -17
Flampunkt: 66
MDL Number: MFCD00000535
CAS nummer: 95-50-1
UN: 1591
ADR: 6.1 III
Merck Index: 13,03081

Order Now

Specifikation testresultat

Purity (GC) min 99%
Free Acid (as HCl) max 0.001%
Chloride (Cl) max 0.0001%
Heavy metals (as Pb) max 0.0001%
Fe (Iron) max 0.0001%
Sulfated ash max 0.001%
Water max 0.01%

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR