n-dekan 95, GPR RECTAPUR®

Leverantör: VWR Chemicals
Fara

87028.320EA 2175 SEK
87028.320 87028.290 87028.230
n-dekan 95, GPR RECTAPUR®
n-dekan
Formel: H₃C(CH₂)₈CH₃
MW: 142.28 g/mol
Kokpunkt: 174.1
Smältpunkt: –30 °C
Densitet: 0.73 g/cm³ (20 °C)
Flampunkt: 46 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008954
CAS nummer: 124-18-5
EINECS: 204-686-4
UN: 2247
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0e4dead3a7841a2258e8a5b99b880895cd06aa1&vid=209e0ebbe6a38426575fcf5608cf6f2b586e6d5e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Assay Min. 95 %
Boiling point Min. 170 °C
Density (20/4) 0,729 --> 0,731
Aromatics Max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR