Krom 1.000 mg/l i utspädd salpetersyra AVS TITRINORM standard för AAS
Leverantör: VWR Chemicals
86671.260EA 545 SEK
86671.260 86671.180
Krom 1.000 mg/l i utspädd salpetersyra AVS TITRINORM standard för AAS
Standarder Cationic Standards Chromium Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%