VWR® PESTINORM®, 1,4-Dioxane, Stabilised, Secondary Reference Standard for GC
Leverantör: VWR Chemicals
85664.180EA 3075 SEK
85664.180
VWR® PESTINORM®, 1,4-Dioxane, Stabilised, Secondary Reference Standard for GC
Standarder Standarder för kromatografi
Stabilised with BHT (2,6-Di-tert-butyl-p-cresol, 2,6-Di-tert-butyl-4-methylphenol, Ionol) 20 ppm. A secondary GC solvent reference standard for various chromatographic and analytical applications.
  • Purity and the specific impurity profile is determined by GC-MS and reported in the CoA
  • Traceability data to specific current lot of primary standard
  • Completely synthetic and typically over 99,9% pure by GC-FID