VWR® PESTINORM®, N,N-Dimethylacetamide, Secondary Reference Standard for GC
Leverantör: VWR Chemicals
85661.180EA 3075 SEK
85661.180
VWR® PESTINORM®, N,N-Dimethylacetamide, Secondary Reference Standard for GC
Standarder Standarder för kromatografi
A secondary GC solvent reference standard for various chromatographic and analytical applications.
  • Purity and the specific impurity profile is determined by GC-MS and reported in the CoA
  • Traceability data to specific current lot of primary standard
  • Completely synthetic and typically over 99,9% pure by GC-FID