VWR®, Colour standards, Saybolt
Leverantör: VWR Chemicals
84799.180EA 1125 SEK
84799.180 84799.260 84800.180 84800.260 84801.180 84801.260 84802.180 84802.260 84803.180 84803.260 84804.180 84804.260 84805.180 84805.260
VWR®, Colour standards, Saybolt
Standarder Färgreferens och spektroskopistandarder
Ready to use colour solutions that are prepared gravimetrically on a weight/weight basis.
  • Use either as calibration and/or quality control standards
  • Designed specifically for use in ASTM analytical methods, and produced in accordance with ASTM D1500, D6045, D1209
  • Traceable product certified by an independent laboratory
  • Supplied in high quality, tamper-evident bottles