VWR® ARISTAR®, Multi-Element Quality Control Standard 10 mg/L, 12 Elements in Nitric Acid 2% / Hydrofluoric Acid < 0.1% for ICP-MS
Leverantör: VWR Chemicals
84794.180EA 3800 SEK
84794.180
VWR® ARISTAR®, Multi-Element Quality Control Standard 10 mg/L, 12 Elements in Nitric Acid 2% / Hydrofluoric Acid < 0.1% for ICP-MS
Standarder Cationic Standards Multi-Element Standards
12 elements at 10 mg/L: Hf, Ge, Mo, Nb, Sb, Si, Sn, Ta, Te, Ti, W, Zr
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
Supplied with certificate of analysis.
Certifikat: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Leveransinformation: Supplied with certificate of analysis.