2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for preparative HPLC

Leverantör: VWR Chemicals
Fara

Synonyms: Isopropanol

Filtered through a 0.2 µm filter, packaged under nitrogen.

84531.550EA 0 SEK
84531.550 84531.460 84531.950
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for preparative HPLC
2-Propanol
Formel: (CH₃)₂CHOH
MW: 60.1 g/mol
Kokpunkt: 82
Smältpunkt: –89 °C
Densitet: 0.786 g/cm³ (20 °C)
Flampunkt: 12
Storage Temperature: Omgivande
MDL Number: MFCD00011674
CAS nummer: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3 II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Assay (on anhydrous substance) Min. 99.8 %
Acidity Max. 0.0005 meq/g
Evaporation residue Max. 5 ppm
Water Max. 0.1 %

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR