Metanol, BAKER ANALYZED®, gradient HPLC/UHPLC, J.T.Baker®

Leverantör: Avantor
Fara

Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

8402.1000EA 435 SEK
8402.1000 8402.2500 8402.5000
Metanol, BAKER ANALYZED®, gradient HPLC/UHPLC, J.T.Baker®
Metanol
Formel: CH₃OH
MW: 32.04 g/mol
Kokpunkt: 64.6
Smältpunkt: –98 °C
Densitet: 0.7918 g/cm³ (20 °C)
Flampunkt: 11 °C (closed cup)
Storage Temperature: Omgivande
MDL Number: MFCD00004595
CAS nummer: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Assay (by GC) (corrected for water) min. 99.8 %
Acetone max. 0.001 %
Residue after Evaporation (in ppm) max. 2
Titrable Acid (meq/g) max. 0.0003
Titrable Base (meq/g) max. 0.0001
Water (H2O) max. 0.02 %
Fluorescence Trace Impurities (as quinine base), ppb:
Measured at Emission Maximum for Solvent Impurities max. 1.0
Measured at 450 nm max. 0.3
Gradient Elution Test Ultraviolet Absorbance (a.u.):
at 235 nm max. 0.005
at 254 nm max. 0.02
Ultraviolet Absorbance (1.00-cm path vs water):
at 225 nm max. 0.20
at 280 nm max. 0.01
at 350 nm max. 0.01
UV Cut-off, nm max. 206


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR