Salpetersyra 67%, NORMATOM® för spårmetallanalys

Leverantör: VWR Chemicals
Fara

Synonyms: Nitric acid solution

83872.270EA 1975 SEK
83872.270 83872.330 83872.290
Salpetersyra 67%, NORMATOM® för spårmetallanalys
Salpetersyra
Formel: HNO₃
MW: 63.01 g/mol
Kokpunkt: 120.5 °C (1013 hPa)
Smältpunkt: –42 °C
Densitet: 1.4 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011349
CAS nummer: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8 (5.1) II
Merck Index: 13,06610

Order Now

Specifikation testresultat

Assay 67 - 69 %
Colouration Max. 10 APHA
Cl (Chloride) Max. 0.2 ppm
Total P (Phosphorus) Max. 0.01 ppm
Total S (Sulphur) Max. 0.3 ppm
Ag (Silver) Max. 0.1 ppb
Al (Aluminium) Max. 1 ppb
As (Arsenic) Max. 0.5 ppb
Au (Gold) Max. 0.1 ppb
B (Boron) Max. 1 ppb
Ba (Barium) Max. 0.1 ppb
Be (Beryllium) Max. 0.1 ppb
Bi (Bismuth) Max. 0.1 ppb
Ca (Calcium) Max. 1 ppb
Cd (Cadmium) Max. 0.5 ppb
Ce (Cerium) Max. 0.1 ppb
Co (Cobalt) Max. 0.5 ppb
Cr (Chromium) Max. 1 ppb
Cs (Cesium) Max. 0.1 ppb
Cu (Copper) Max. 0.5 ppb
Dy (Dysprosium) Max. 0.1 ppb
Er (Erbium) Max. 0.1 ppb
Eu (Europium) Max. 0.1 ppb
Fe (Iron) Max. 1 ppb
Ga (Gallium) Max. 0.1 ppb
Gd (Gadolinium) Max. 0.1 ppb
Ge (Germanium) Max. 0.1 ppb
Hf (Hafnium) Max. 0.1 ppb
Hg (Mercury) Max. 0.1 ppb
Ho (Holmium) Max. 0.1 ppb
In (Indium) Max. 0.1 ppb
K (Potassium) Max. 1 ppb
La (Lanthanum) Max. 0.1 ppb
Li (Lithium) Max. 0.1 ppb
Lu (Lutetium) Max. 0.1 ppb
Mg (Magnesium) Max. 1 ppb
Mn (Manganese) Max. 0.1 ppb
Mo (Molybdenum) Max. 0.1 ppb
Na (Sodium) Max. 1 ppb
Nb (Niobium) Max. 0.1 ppb
Nd (Neodymium) Max. 0.1 ppb
Ni (Nickel) Max. 0.5 ppb
Pb (Lead) Max. 0.1 ppb
Pd (Palladium) Max. 0.5 ppb
Pr (Praseodymium) Max. 0.1 ppb
Pt (Platinum) Max. 0.5 ppb
Rb (Rubidium) Max. 0.1 ppb
Re (Rhenium) Max. 0.1 ppb
Rh (Rhodium) Max. 0.5 ppb
Ru (Ruthenium) Max. 0.500 ppb
Sb (Antimony) Max. 0.500 ppb
Sc (Scandium) Max. 0.100 ppb
Se (Selenium) Max. 1 ppb
Sm (Samarium) Max. 0.100 ppb
Sn (Stagno) Max. 0,500 ppb
Sr (Strontium) Max. 0.100 ppb
Tb (Terbium) Max. 0.100 ppb
Te (Tellurium) Max. 0.100 ppb
Th (Thorium) Max. 0.100 ppb
Ti (Titanium) Max. 0.500 ppb
Tl (Thallium) Max. 0.100 ppb
Tm (Thulium) Max. 0.100 ppb
U (Uranium) Max. 0.100 ppb
V (Vanadium) Max. 0.500 ppb
W (Tungsten) Max. 0.100 ppb
Y (Yttrium) Max. 0.100 ppb
Yb (Ytterbium) Max. 0.100 ppb
Zn (Zinc) Max. 0.500 ppb
Zr (Zirconium) Max. 0.100 ppb


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR