Aceton, vattenfri (högst 0,0100% H₂O) ≥99.8%

Leverantör: VWR Chemicals
Fara

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

Filtered through a 0.2 µm filter, packaged under nitrogen.

83683.230EA 995 SEK
83683.230 83683.290 83683.269
Aceton, vattenfri (högst 0,0100% H₂O) ≥99.8%
Aceton

250 ml bottle with a septum cap featuring six separate re-sealable puncture points


250 and 500 ml bottles with a septum cap featuring six separate re-sealable puncture points

Formel: CH₃COCH₃
MW: 58.08 g/mol
Kokpunkt: 56.2 °C (1013 hPa)
Smältpunkt: –95.4 °C
Densitet: 0.792 g/cm³ (20 °C)
Flampunkt: <–20 °C (closed cup)
Storage Temperature: Omgivande
MDL Number: MFCD00008765
CAS nummer: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3 II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Purity (GC) Min. 99.8 %
Acidity Max. 0.0005 meq/g
Residue on evaporation Max. 0.0005 %
Water Max. 0.0100 %

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR