Metanol, vattenfri (högst 0,0020% H2O) ≥99.9%

Leverantör: VWR Chemicals
Fara

Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

Filtered through a 0.2 µm filter, packaged under nitrogen.

83679.230EA 860 SEK
83679.230 83679.290 83679.260
Metanol, vattenfri (högst 0,0020% H2O) ≥99.9%
Metanol

250 ml bottle with a septum cap featuring six separate re-sealable puncture points

Formel: CH₃OH
MW: 32.04 g/mol
Kokpunkt: 64.6 °C (1013 hPa)
Smältpunkt: –98 °C
Densitet: 0.7918 g/cm³ (20 °C)
Flampunkt: 11 °C (closed cup)
Storage Temperature: Omgivande
MDL Number: MFCD00004595
CAS nummer: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 (6.1) II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Assay Min. 99.9 %
Acidity Max. 0.00050 meq/g
Residue on evaporation Max. 0.0003 %
Water (K.F.) Max. 0.0020 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR