Propylbenzen, Supelco®

Leverantör: Merck
Fara


M-Clarity™ quality level = MQ100

82118-10ML 82118-5ML
82118-10MLEA 1710 SEK
82118-10ML 82118-5ML
Propylbenzen, Supelco®
Propylbenzen
Formel: C₉H₁₂
MW: 120.19 g/mol
Kokpunkt: 159
Smältpunkt: –99 °C
Densitet: 0.862 g/cm³ (20 °C)
Flampunkt: 45
Storage Temperature: Omgivande
MDL Number: MFCD00009377
CAS nummer: 103-65-1
EINECS: 203-132-9
UN: 2364
ADR: 3 III
Merck Index: 12,08029
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a5ddcc46e456613b0b0417e84289f17ae683889d&vid=299912d529669903728d8645724c7f1a0c956315&prid=web&clid=SAPDAM

Order Now

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR