3,5-Dikloranilin zS, Sigma-Aldrich®

Leverantör: Merck
Fara


MQ200

8.41790.0100
8.41790.0100EA 1670 SEK
8.41790.0100
3,5-Dikloranilin zS, Sigma-Aldrich®
3,5-Dikloranilin
Formel: C₆H₅Cl₂N
MW: 162.02 g/mol
Kokpunkt: 262
Smältpunkt: 51
Densitet: 1.58 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00007774
CAS nummer: 626-43-7
EINECS: 210-948-9
UN: 3442
ADR: 6.1 II

Order Now

Specifikation testresultat

Solubility 0.78 g/l
Melting Point 46 - 53 °C
Molar Mass 162.02 g/mol
Boiling Point 259 - 260 °C (1013 hPa)
Vapor Pressure 40 hPa (150 °C)
Flash Point 133 °C
Density 1.58 g/cm3 (20 °C)
Ignition Point 620 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR