Cholesteryl chloroformate zS

Leverantör: Merck

8.41499.0025
8.41499.0025EA 0 SEK
8.41499.0025
Cholesteryl chloroformate zS
Cholesteryl chloroformate
Formel: C₂₈H₄₅ClO₂
MW: 449.12 g/mol
MDL Number: MFCD00003633
CAS nummer: 7144-08-3
EINECS: 230-447-9
UN: 3261
ADR: 8 II

Order Now

Specifikation testresultat

Melting Point 117 - 119 °C
Molar Mass 449.11 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR