3,5-Dichloro-2-hydroxybenzenesulphonic acid sodium salt för syntes, Sigma-Aldrich®

Leverantör: Merck
Varning

Synonyms: Sodium 3,5-Dichloro-2-hydroxybenzenesulphonate, DHBS

M-Clarity™ quality level = MQ200

8.41262.0005 8.41262.0025
8.41262.0005EA 1210 SEK
8.41262.0005 8.41262.0025
3,5-Dichloro-2-hydroxybenzenesulphonic acid sodium salt för syntes, Sigma-Aldrich®
3,5-Dichloro-2-hydroxybenzenesulphonic acid sodium salt
Formel: C₆H₃Cl₂NaO₄S
MW: 265.05 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00009798
CAS nummer: 54970-72-8
EINECS: 259-416-8

Order Now

Specifikation testresultat

Solubility (20 °C) soluble
Melting Point 300 °C
Molar Mass 265.05 g/mol
pH 7 (53 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR