Pentafluoropropionic acid zS, Sigma-Aldrich®

Leverantör: Merck
Fara

Synonyms: Perfluoropropionic acid, Pentafluoropropanoic acid, Perfluoropropanoic acid, 2,2,3,3,3-Pentafluoropropionic acid

MQ200

8.20956.0010 8.20956.0050
8.20956.0010EA 1230 SEK
8.20956.0010 8.20956.0050
Pentafluoropropionic acid zS, Sigma-Aldrich®
Pentafluoropropionic acid
Formel: C₃HF₅O₂
MW: 164.03 g/mol
Kokpunkt: 96
Densitet: 1.561 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00004170
CAS nummer: 422-64-0
EINECS: 207-021-6
UN: 2922
ADR: 8 (6.1) II

Order Now

Specifikation testresultat

Solubility (20 °C) soluble
Molar Mass 164.03 g/mol
Boiling Point 96 - 97 °C (984 hPa)
Vapor Pressure 52 hPa (25 °C)
Refractive Index 1.2838 (25 °C, 589 nm)
Density 1.57 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR