Norbornadiene stabilised för syntes, Sigma-Aldrich®

Leverantör: Merck
Fara

Synonyms: 2,5-Norbornadiene

M-Clarity™ quality level = MQ200

8.20918.0250
8.20918.0250EA 2920 SEK
8.20918.0250
Norbornadiene stabilised för syntes, Sigma-Aldrich®
Norbornadiene
Formel: C₇H₈
MW: 92.14 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00082301
CAS nummer: 121-46-0
EINECS: 204-472-0
UN: 2251
ADR: 3 II

Order Now

Specifikation testresultat

Solubility (20 °C) practically insoluble
Melting Point -20 °C
Molar Mass 92.14 g/mol
Boiling Point 90 °C
Vapor Pressure 69 hPa (20 °C)
Flash Point -11 °C
Refractive Index 1.4702 (20 °C, 589 nm)
Explosion Limit 1.6 - 6.3 %(V)
Density 0.906 g/cm3 (20 °C)
Ignition Point 350 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR