Naftalen zS, Sigma-Aldrich®

Leverantör: Merck
Varning


MQ200

8.20846.0100 8.20846.1000 8.20846.9010
8.20846.0100EA 425 SEK
8.20846.0100 8.20846.1000 8.20846.9010
Naftalen zS, Sigma-Aldrich®
Naftalen
Formel: C₁₀H₈
MW: 128.17 g/mol
Kokpunkt: 218
Smältpunkt: 79…82 °C
Densitet: 1.145 g/cm³ (20 °C)
Flampunkt: 79
Storage Temperature: Omgivande
MDL Number: MFCD00001742
CAS nummer: 91-20-3
EINECS: 202-049-5
UN: 1334
ADR: 4.1 III
Merck Index: 12,06457
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c87c606cb828e0f39bab8b9d50c2c26501e1637d&vid=9e4bc21353014391fc8486208c8757e6375e2ef5&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Solubility 0.03 g/l (25 °C)
Melting Point 79 - 82 °C
Molar Mass 128.17 g/mol
Bulk Density 550 - 600 mg/m3
Boiling Point 218 °C
Vapor Pressure 0.08 hPa (20 °C)
Refractive Index 80 °C
Explosion Limit 0.9 - 5.9 %(V)
Density 1.15 g/cm3 (20 °C)
Ignition Point 540 °C
Hazard Class 4.1
Hazard Class Description Flammable Solid

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR