N-(3-sulphopropyl)-N-methacroyloxyethyl-N,N-dimethylammonium betaine zS, Sigma-Aldrich®

Leverantör: Merck

Synonyms: N,N-Dimethyl-N-(2-methacryloxyethyl)-N-(3-sulphopropyl)ammonium betaine, 3-((2-(Methacryloyloxy)ethyl)dimethylammonio)propane-1-sulphonate, 3-[[2-(Methacryloyloxy)ethyl]dimethylammonio]propane-1-sulphonate, SPE, DMAPS, [2-(Methacryloyloxy)ethyl]dimethyl(3-sulphopropyl)ammonium Hydroxide Inner salt

MQ200

8.18484.0100
8.18484.0100EA 14525 SEK
8.18484.0100
N-(3-sulphopropyl)-N-methacroyloxyethyl-N,N-dimethylammonium betaine zS, Sigma-Aldrich®
N-(3-sulphopropyl)-N-methacroyloxyethyl-N,N-dimethylammonium betaine
Formel: C₁₁H₂₁NO₅S
MW: 279.351 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00040574
CAS nummer: 3637-26-1
EINECS: 222-860-8

Order Now

Specifikation testresultat

Hill Formula C₁₁H₂₁NO₅S
Molar Mass 279.36 g/mol
Flash point 110 °C
Melting Point 147 - 149 °C
pH value 6.3 (100 g/l, H₂O, 20 °C)
Bulk density 400 kg/m³
Solubility 500 g/l (20 °C)
Storage class 10 - 13 Other liquids and solids
WGK WGK 3 highly water endangering


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR