Polyetylenglykol 3.000, EMPROVE® ESSENTIAL Ph. Eur. suitable for use as excipient, SAFC®

Leverantör: MERCK PRODUCTION CHEMICALS

Synonyms: PEG 3,000

8.17019.5000
8.17019.5000EA 6300 SEK
8.17019.5000
Polyetylenglykol 3.000, EMPROVE® ESSENTIAL Ph. Eur. suitable for use as excipient, SAFC®
Polyetylenglykol 3.000
Formel: H(OCH₂CH₂)nOH
MW: 3000 g/mol
Smältpunkt: 55…58 °C
Densitet: 1.21 g/cm³ (20 °C)
Storage Temperature: Omgivande
CAS nummer: 25322-68-3
EINECS: 500-038-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR