n-Tridekan zS, Sigma-Aldrich®

Leverantör: Merck
Fara

Synonyms: Tridecane

MQ200

8.14631.0025
8.14631.0025EA 467 SEK
8.14631.0025
n-Tridekan zS, Sigma-Aldrich®
n-Tridekan
Formel: H₃C(CH₂)₁₁CH₃
MW: 184.37 g/mol
Kokpunkt: 235.4
Smältpunkt: –5.5 °C
Densitet: 0.7564 g/cm³ (20 °C)
Flampunkt: 79
Storage Temperature: Omgivande
MDL Number: MFCD00008979
CAS nummer: 629-50-5
EINECS: 211-093-4
UN: 3295

Order Now

Specifikation testresultat

Solubility (25 °C) practically insoluble
Melting Point -5 °C
Molar Mass 184.36 g/mol
Boiling Point 235 °C (1013 hPa)
Vapor Pressure  - 0.5 hPa (20 °C)
Flash Point 79 °C
Explosion Limit 0.6 - 6.5 %(V)
Density 0.757 g/cm3 (20 °C)
Ignition Point 204 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR