2-Ethylpyrazine zS

Leverantör: Merck

Synonyms: Ethylpyrazine

8.14478.0005
8.14478.0005EA 0 SEK
8.14478.0005
2-Ethylpyrazine zS
2-Ethylpyrazine
Formel: C₆H₈N₂
MW: 108.14 g/mol
Kokpunkt: 152
Densitet: 0.98 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00006149
CAS nummer: 13925-00-3
EINECS: 237-691-5
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR