Lithium tetrafluoroborate zS, Sigma-Aldrich®

Leverantör: Merck
Fara


MQ200

8.14347.0010
8.14347.0010EA 840 SEK
8.14347.0010
Lithium tetrafluoroborate zS, Sigma-Aldrich®
Lithium tetrafluoroborate
Formel: BF₄Li
MW: 93.74 g/mol
Densitet: 0.852 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011087
CAS nummer: 14283-07-9
EINECS: 238-178-9
UN: 3260
ADR: 8 II

Order Now

Specifikation testresultat

Solubility (20 °C) soluble
Melting Point 324 °C (decomposition)
Molar Mass 93.74 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR