Bis(4-hydroxyphenyl)sulphide zS, Sigma-Aldrich®

Leverantör: Merck
Varning

Synonyms: 4,4'-Thiodiphenol

MQ200

8.14209.0100
8.14209.0100EA 325 SEK
8.14209.0100
Bis(4-hydroxyphenyl)sulphide zS, Sigma-Aldrich®
Bis(4-hydroxyphenyl)sulphide
Formel: C₁₂H₁₀O₂S
MW: 218.28 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00002349
CAS nummer: 2664-63-3
EINECS: 220-197-9
UN: 3261

Order Now

Specifikation testresultat

Solubility (20 °C) insoluble
Melting Point 149 - 152 °C
Molar Mass 218.26 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR