Zirconium(IV) acetylacetonate, Sigma-Aldrich®

Leverantör: Merck
Varning

Synonyms: Tetrakis(2,4-pentanedionato)zirconium(IV), Zirconium(IV) 2,4-pentanedionate

MQ200

8.08915.0100
8.08915.0100EA 932 SEK
8.08915.0100
Zirconium(IV) acetylacetonate, Sigma-Aldrich®
Zirconium(IV) acetylacetonate
Formel: C₂₀H₂₈O₈Zr
MW: 487.66 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00000036
CAS nummer: 17501-44-9
EINECS: 241-510-5

Order Now

Specifikation testresultat

Solubility 14 g/l (20 °C)
Melting Point 189 - 192 °C
Molar Mass 487.66 g/mol
Bulk Density 500 kg/m3
pH 7.2 (4.5 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR