Ethyl tridecanoate zS, Sigma-Aldrich®

Leverantör: Merck

Synonyms: Tridecylic acid ethyl ester

MQ200

8.08634.0010
8.08634.0010EA 1220 SEK
8.08634.0010
Ethyl tridecanoate zS, Sigma-Aldrich®
Ethyl tridecanoate
Formel: C₁₅H₃₀O₂
MW: 242.4 g/mol
Densitet: 0.86 g/cm³ (20 °C)
MDL Number: MFCD00008978
CAS nummer: 28267-29-0
EINECS: 248-929-2

Order Now

Specifikation testresultat

Solubility (20 °C) insoluble
Molar Mass 242.4 g/mol
Density 0.86 g/cm3 (20 °C)
Synonyms Tridecanoic acid ethyl ester
Hill Formula C15H30O2
Chemical Formula CH3(CH2)11COOC2H5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR