Ethyl toluene-4-sulphonate för syntes, Sigma-Aldrich®

Leverantör: Merck

Synonyms: Ethyl 4-toluenesulphonate, Ethyl p-toluenesulphonate, p-Toluenesulphonic acid ethyl ester, Ethyl 4-methylbenzenesulphonate, Toluene-4-sulphonic acid ethyl ester

M-Clarity™ quality level = MQ200

8.08332.0100 8.08332.0500 8.08332.5000
8.08332.0100EA 400 SEK
8.08332.0100 8.08332.0500 8.08332.5000
Ethyl toluene-4-sulphonate för syntes, Sigma-Aldrich®
Ethyl toluene-4-sulphonate
Formel: C₉H₁₂O₃S
MW: 200.26 g/mol
Kokpunkt: 158
Smältpunkt: 30…32 °C
Densitet: 1.174 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00009100
CAS nummer: 80-40-0
EINECS: 201-276-7
UN: 2811
Merck Index: 13,03889

Order Now

Specifikation testresultat

Solubility 1.24 g/l (25 °C)
Melting Point 30 - 32 °C
Molar Mass 200.25 g/mol
Boiling Point 221 °C (1013 hPa)
Vapor Pressure 0.6 hPa (14 °C)
Flash Point 158 °C
Density 1.174 g/cm3 (20 °C)
Ignition Point 500 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR