(±)-Ethyl mandelate, Sigma-Aldrich®

Leverantör: Merck
Varning

Synonyms: Ethyl 2-hydroxy-2-phenylacetate

MQ200

8.06912.0100
8.06912.0100EA 690 SEK
8.06912.0100
(±)-Ethyl mandelate, Sigma-Aldrich®
(±)-Ethyl mandelate
Formel: C₁₀H₁₂O₃
MW: 180.2 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00004494
CAS nummer: 774-40-3
EINECS: 212-263-0

Order Now

Specifikation testresultat

Solubility (20 °C) insoluble
Melting Point 24 - 27 °C
Molar Mass 180.2 g/mol
Boiling Point 253 - 255 °C
Flash Point 93 °C
Refractive Index 1.5136 (20 °C, 589 nm)
Density 1.13 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR