4-Klorbenzaldehyd zS, Sigma-Aldrich®

Leverantör: Merck
Varning


MQ200

8.02453.0250 8.02453.1000 8.02453.0005
8.02453.0250EA 524 SEK
8.02453.0250 8.02453.1000 8.02453.0005
4-Klorbenzaldehyd zS, Sigma-Aldrich®
4-Klorbenzaldehyd
Formel: C₇H₅ClO
MW: 140.57 g/mol
Kokpunkt: 214
Smältpunkt: 46
Densitet: 1.196 g/cm³ (20 °C)
Flampunkt: 88
Storage Temperature: Omgivande
MDL Number: MFCD00003379
CAS nummer: 104-88-1
EINECS: 203-247-4
UN: 3077
ADR: 9 III

Order Now

Specifikation testresultat

Solubility 935 mg/l (20 °C)
Melting Point 46 °C
Molar Mass 140.57 g/mol
Boiling Point 214 °C (1013 hPa)
Vapor Pressure 0.36 hPa (25 °C)
Flash Point 88 °C
Density 1.19 g/cm3 (60 °C)
Ignition Point 302 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR