Ethyl benzoylacetate, Sigma-Aldrich®

Leverantör: Merck

Synonyms: Ethyl 3-oxo-3-phenylpropanoate

M-Clarity™ quality level = MQ200

8.01808.0100 8.01808.0250 8.01808.9010
8.01808.0100EA 629 SEK
8.01808.0100 8.01808.0250 8.01808.9010
Ethyl benzoylacetate, Sigma-Aldrich®
Ethyl benzoylacetate
Formel: C₁₁H₁₂O₃
MW: 192.21 g/mol
Kokpunkt: 265
Smältpunkt: < 0 °C
Densitet: 1.114 g/cm³ (20 °C)
Flampunkt: 140
MDL Number: MFCD00009196
CAS nummer: 94-02-0
EINECS: 202-295-3
Merck Index: 13,03803

Order Now

Specifikation testresultat

Solubility (20 °C) insoluble
Molar Mass 192.21 g/mol
Boiling Point 265 - 270 °C (decomposition)
Vapor Pressure 1 hPa (108 °C)
Flash Point 140 °C
Refractive Index 1.531 (20 °C, 589 nm)
Density 1.114 g/cm3 (20 °C)
Ignition Point 375 °C DIN 51794
Synonyms Benzoylacetic acid ethyl ester, 3-Oxo-3-phenylpropionic acid ethyl ester
Hill Formula C11H12O3
Chemical Formula C6H5COCH2COOC2H5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR