N-Etyldiisopropylamin zS, Sigma-Aldrich®

Leverantör: Merck
Fara

Synonyms: Hünig’s base, N,N-Diisopropylethylamine, N.N’-Diisopropyl ethylamine, DIPEA

MQ200

8.00894.0250 8.00894.1000 8.00894.9150
8.00894.0250EA 1320 SEK
8.00894.0250 8.00894.1000 8.00894.9150
N-Etyldiisopropylamin zS, Sigma-Aldrich®
N-Etyldiisopropylamin
Formel: C₈H₁₉N
MW: 129.25 g/mol
Kokpunkt: 127
Smältpunkt: –50 °C
Densitet: 0.76 g/cm³ (20 °C)
Flampunkt: 6
Storage Temperature: Omgivande
MDL Number: MFCD00008868
CAS nummer: 7087-68-5
EINECS: 230-392-0
UN: 1992
ADR: 3 (6.1) II

Order Now

Specifikation testresultat

Solubility 3.9 g/l (20°C)
Melting Point -46 °C
Boiling Point 127 °C (1013 hPa)
Vapor Pressure 16 hPa (20 °C)
Flash Point 9.5 °C
Refractive Index 1.4121 (25 °C, 589 nm)
Explosion Limit 0.7 - 6.3 %(V)
Density 0.76 g/cm3 (20 °C)
pH 12.3 (H2O) (as an emulsion)
Ignition Point 240°C
Viscosity Kinematic 0.88 mm2/s (20 °C)
Synonyms N,N-Diisopropylethylamine
Hill Formula C8H19N
Chemical Formula [(CH3)2CH]2NCH2CH3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR