Adiponitrile, Sigma-Aldrich®

Leverantör: Merck
Fara

Synonyms: Hexanedinitrile

M-Clarity™ quality level = MQ200

8.00092.0250
8.00092.0250EA 615 SEK
8.00092.0250
Adiponitrile, Sigma-Aldrich®
Adiponitrile
Formel: C₆H₈N₂
MW: 108.14 g/mol
Kokpunkt: 296
Smältpunkt: 2
Densitet: 0.965 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00001975
CAS nummer: 111-69-3
EINECS: 203-896-3
UN: 2205
ADR: 6.1 III

Order Now

Specifikation testresultat

Solubility 50 g/l (20 °C)
Melting Point 2 °C
Molar Mass 108.14 g/mol
Boiling Point 295 °C
Vapor Pressure 0.003 hPa (20 °C)
Flash Point 93 °C (decomposition)
Refractive Index 1.438 (20 °C, 589 nm)
Explosion Limit 7 - 14 %(V)
Density 0.96 g/cm3 (20 °C)
Ignition Point 460 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR