Bovine aprotinin
Leverantör: G Biosciences
786-046
786-046EA 11225 SEK
786-046
Bovine aprotinin
Proteiner
Aprotinin is a broad range, competitive and reversible inhibitor of chymotrypsin, plasmin, trypsin, kallikrein and other serine proteases. Activity: approx 6,000 KIU/mg.