Handvakuumpump med pumptallrik
Leverantör: CORNELSEN EXPERIMENTA
45330
765-0144EA 0 SEK
765-0144
Handvakuumpump med pumptallrik
Pumpar Handvakuumpumpar
Piston air pump on a stable plastic base with rubber feet, non-return valve with a bleed screw, pump plate and vaccumm connection to the centre of the plate.
  • Plate diameter: 240 mm
  • Capable of 50 mbar pressure