Crystallisation plates, CrystalStar™, 1536-well IMP@CT™
Leverantör: Greiner Bio-One
656190_200
737-3507EA 1725 SEK
737-3507
Crystallisation plates, CrystalStar™, 1536-well IMP@CT™
Plattor Kristallisationsplattor
Designed for microbatch applications under oil, the IMP@CT™ crystallisation plates are characterised by conical wells with a flat bottom.