96-well polyvinyl chloride (PVC) microplates
Leverantör: Corning
2797
734-1517EA 2925 SEK
734-1517
96-well polyvinyl chloride (PVC) microplates
Plattor Cellodlingsplattor
Not treated PVC microplates are economical microplates for solution-based assays, serial dilutions, and general storage applications.
  • Well volume of 250 µl (260 µl for V-bottom)
  • Working well volume of 50 µl to 150 µl
Beställningsinformation: Lids are not available.