Collagen I-coated flasks, Corning® BioCoat®
Leverantör: Corning
354484 354485 354486 354487 356484 356485 356486 356487
734-0158EA 1850 SEK
734-0158 734-0159 734-0160 734-0161 734-0289 734-0290 734-0291 734-0292
Collagen I-coated flasks, Corning® BioCoat®
Kolvar Cellodlingsflaskor
Collagen I, found in most tissues and organs, is most plentiful in dermis, tendon, and bone. It is an integral part of the framework that holds cells and tissues together and has been recognized as a useful matrix for improving cell culture. In vitro use of collagen can exert effects on the adherence, morphology, growth, migration, and differentiation of a variety of cell types.
  • Promotion of cell attachment and spreading
  • Rapid expansion of cell populations
  • Serum-free or reduced serum culture
  • Cell adhesion assays
  • Improving survival of primary cells in
More Product Information