TLC imaging system, MultiDoc-It™
Leverantör: UVP ULTRA VIOLET PRODUCTS
97-0202-02
733-2048EA 0 SEK
733-2048
TLC imaging system, MultiDoc-It™
Fotodokumentationssystem
For imaging in thin layer chromatography (TLC) applications, with mega pixel resolution digital colour camera for capturing gels, plates and membranes.
  • Sturdy metal construction with large access door
  • Hood enclosure provides a darkroom environment and protects the Digi camera
  • Exclusive capture software enables image acquisition, enhancement and report capabilities
  • Built-in white light; safety switch turns UV off when the door is open
  • W×D×H: 356×323×483 mm
Leveransinformation: MultiDoc-It™ TLC model includes two UVGL-25 UV lamps for placement on twoMore Product Information
Order Now
SPECIFIKATIONER
Environmentally Preferable