Electroporation system, twin-wave, Gemini X² and SC
Leverantör: BTX
45-2044 45-2040 45-2041 45-2042 45-2043
732-3775EA 0 SEK
732-3775 732-3771 732-3772 732-3773 732-3774
Electroporation system, twin-wave, Gemini X² and SC
Elektroporeringssystem Elektroporatorer
Gemini twin-wave electroporators are flexible systems allowing both square wave and exponential decay wave electroporation in a single unit. These waveform combinations enable researchers to easily and efficiently electroporate eukaryotic and prokaryotic cells in cuvettes or plates with one easy to use set-up. Ideal for all the electroporation needs, including CRISPR, in vivo, in vitro, in ovo and more.
  • Universal electroporation – transfects cells in vitro, in vivo, in ovo and adherent forms
  • Multiple pulsing capabilities in both square and exponential
More Product Information