Handhållna UV-lampor
Leverantör: UVP ULTRA VIOLET PRODUCTS
95-0104-02 95-0006-03 95-0004-10 95-0007-06 95-0005-06
730-1024EA 7025 SEK
730-1024 733-2421 732-1109 732-0857 733-2365
Handhållna UV-lampor
Belysning UV-lampor
These lightweight lamps can be used with the optional lamp stand for hands-free operation. Replacement tubes are also available.
  • Lightweight for easy handling
  • Durable plastic housing
  • Six watts
  • Models available with short wave, long wave UV or a combination short wave/long wave version
  • Combination short wave/long wave version comes with a snap-on wavelength selector for switching between wavelengths (UVGL-58 only)
L×B×H: 378×81×64 mm
Vikt: 0,9 kg