Gel analysis software, Dymension
Leverantör: Syngene (a division of the Synoptics group)
DYM-1 DYM-2 DYM-3
730-1161EA 19425 SEK
730-1161 730-1162 730-1163
Gel analysis software, Dymension
Software Laboratory Software
Dymension software can be used to analyse a typical 2D gel image rapidly. It features algorithms for background subtraction, noise filtering, precise alignment, spot detection, rapid matching and reduced image editing time. Using its powerful spot detection algorithm, Dymension instantly locates and analyses protein spots. With Dymension, the entire analysis process from background correction to spot matching results and reporting takes minutes. Dymension is available in three versions, and upgrading between the packages is simple.
Dymension 1 is ideal for users looking for software toMore Product Information